accessibility menu, dialog, popup

UserName

CAS RN 12190-79-3

IUPAC Name: lithium(1+) cobaltoylolate
Synonyms: lithium cobalt oxide
Molecular Weight (g/mol): 97.87
Molecular Formula: CoLiO2
InChi Key: BFZPBUKRYWOWDV-UHFFFAOYSA-N
SMILES: [Li+].[O-][Co]=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)