missing translation for 'onlineSavingsMsg'
Learn More

Lithium Cobaltite, LiCoO2, MP Biomedicals

Product Code. 11438465 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438465 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438465 Supplier MP Biomedicals Supplier No. 0522108580

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Molecular formula: CoLiO2
  • Molecular weight: 97.871 g/mol
  • CAS number: 12190-79-3
  • Synonym: Lithium cobaltite

Specifications

CAS 12190-79-3
Quantity 100 g
Molecular Formula CoLiO2
Linear Formula LiCoO2
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
Solubility Information Insoluble in water
InChI Formula InChI=1S/Co.Li.2O/q;+1;;-1
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
SMILES [Li+].[O-][Co]=O
IUPAC Name lithium;oxido(oxo)cobalt
Molecular Weight (g/mol) 97.871
PubChem CID 23670860
Formula Weight 97.871
Appearance Gray Powder
Chemical Name or Material Lithium Cobaltite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • LITHIUM COBALTITE, LICOO2, 100 G
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Lithium cobalt(III)oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.