accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 122-79-2

IUPAC Name: phenyl acetate
Synonyms: phenyl acetate acetic acid,phenyl ester fenylester kyseliny octove acetylphenol
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: IPBVNPXQWQGGJP-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)