accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 122-94-1

IUPAC Name: 4-butoxyphenol
Synonyms: 4-butoxyphenol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: MBGGFXOXUIDRJD-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results