accessibility menu, dialog, popup

UserName

OOHFF

CAS RN 122665-97-8

IUPAC Name: 4,4-difluorocyclohexane-1-carboxylic acid
Synonyms: 4,4-difluorocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 164.15
Molecular Formula: C7H10F2O2
InChi Key: HYIUDFLDFSIXTR-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(F)(F)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results