missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11485550
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485550 1 g 1g
15493307 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485550 Supplier Thermo Scientific Alfa Aesar Supplier No. H25831.03
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122665-97-8 | Molecular Formula: C7H10F2O2 | Molecular Weight: 164.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122665-97-8
Molecular Formula C7H10F2O2
Molecular Weight (g/mol) 164.152
MDL Number MFCD03788493
InChI Key HYIUDFLDFSIXTR-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
PubChem CID 2779136
IUPAC Name 4,4-difluorocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)(F)F

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Formula Weight 164.15
Percent Purity 98%
Chemical Name or Material 4,4-Difluorocyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.