accessibility menu, dialog, popup

UserName

H3COOCH3BOHHO

CAS RN 122775-35-3

IUPAC Name: (3,4-dimethoxyphenyl)boronic acid
Synonyms: 3,4-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: RCVDPBFUMYUKPB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)B(O)O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results