missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzeneboronic acid, 98%

Product Code. 11421287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421287 25 g 25g
11401287 1 g 1g
11411287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421287 Supplier Thermo Scientific Alfa Aesar Supplier No. B24240.14
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122775-35-3 | C8H11BO4 | 181.98 g/mol

3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (25 g/L).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122775-35-3
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.98
MDL Number MFCD01074574
InChI Key RCVDPBFUMYUKPB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenyl boronic acid, 3,4-dimethoxybenzeneboronic acid, 3,4-dimethoxyphenyl boranediol, 3,4-dimethoxybenzene boronic acid, boronic acid, 3,4-dimethoxyphenyl, 3,4-dimethoxyphenylboronicacid, 3, 4-dimethoxyphenylboronic acid, pubchem1826, acmc-209amm, ksc174q5t
PubChem CID 2734702
IUPAC Name (3,4-dimethoxyphenyl)boronic acid
SMILES COC1=CC=C(C=C1OC)B(O)O

Specifications

Melting Point ∼240°C
Quantity 25 g
Beilstein 5334877
Solubility Information Soluble in water (25g/L).
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.