CAS RN 1228837-05-5
CAS RN 1228837-05-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(4-Chlorophenyl)-4,4-dimethyl-1-cyclohexene-1-carboxaldehyde, TRC
CAS: 1228837-05-5 Molecular Formula: C15H17ClO Molecular Weight (g/mol): 248.75 Synonym: 2-(4-Chlorophenyl)-4,4-dimethylcyclohex-1-ene-1-carboxaldehyde,2-(4-Chlorophenyl)-4,4-dimethylcyclohex-1-enecarboxaldehyde IUPAC Name: 2-(4-chlorophenyl)-4,4-dimethylcyclohexene-1-carbaldehyde SMILES: CC1(C)CCC(=C(C1)c2ccc(Cl)cc2)C=O