accessibility menu, dialog, popup

UserName

CAS RN 123-33-1

ONHNHO

CAS RN 123-33-1

IUPAC Name: 1,2,3,6-tetrahydropyridazine-3,6-dione
Synonyms: maleic hydrazide stuntman
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: BGRDGMRNKXEXQD-UHFFFAOYSA-N
SMILES: O=C1NNC(=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results