missing translation for 'onlineSavingsMsg'
Learn More

Maleic hydrazide, 97%, Thermo Scientific™

Product Code. 11484194
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484194 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484194 Supplier Thermo Scientific Alfa Aesar Supplier No. A12531.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Maleic hydrazide is a synthetic plant growth regulator that is widely used for controlling growth and to induce dormancy in some crops.

Solubility
Soluble in water (4.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-33-1
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00006665
InChI Key BGRDGMRNKXEXQD-UHFFFAOYSA-N
Synonym pyridazine-3,6-diol, maleic hydrazide, 3,6-dihydroxypyridazine, 3,6-pyridazinediol, stuntman, maleic acid hydrazide, vondalhyde, 3,6-pyridazinedione, 1,2-dihydro, 1,2-dihydro-3,6-pyridazinedione, antergon
PubChem CID 21954
ChEBI CHEBI:81771
IUPAC Name 1,2-dihydropyridazine-3,6-dione
SMILES C1=CC(=O)NNC1=O

Specifications

Melting Point ∼305°C (decomposition)
Density 1.6
Flash Point 300°C (572°F)
Quantity 100 g
Beilstein 112223
Merck Index 14,5705
Solubility Information Soluble in water (4.5g/L).
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material Maleic hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.