accessibility menu, dialog, popup

UserName

CAS RN 123333-68-6

IUPAC Name: 2-sulfobenzoic acid hydrate
Synonyms: benzoic acid, 2-sulfo- hydrate
Molecular Weight (g/mol): 220.2
Molecular Formula: C7H8O6S
InChi Key: HVJWSZOEEIYKSS-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC=CC=C1S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results