CAS RN 1233533-04-4
CAS RN 1233533-04-4
IUPAC Name:
4-hydroxy-8,14-dithia-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Synonyms:
4-hydroxy-8,14-dithia-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Molecular Weight (g/mol):
251.32
Molecular Formula:
C11H9NO2S2
InChi Key:
DHUAGGSHTKPOHU-UHFFFAOYSA-N
SMILES:
OC1=CC=C2SC3=C(SCCNC3=O)C2=C1
1
–
2
of
2
results
1
Tocris Bioscience™ kb NB 142-70
CAS: 1233533-04-4 Molecular Formula: C11H9NO2S2
2
kb NB 142-70, MedChemExpress
MedChemExpress kb NB 142-70 is a potent PKD inhibitor, with IC50s of 28.3, 58.7 and 53.2 nM for PKD1, PKD2, and PKD3, respectively. kb NB 142-70 also has antitumor activity.