CAS RN 1233921-75-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2’,2’-Difluoro-2’-deoxyuridine-13C,15N2, TRC
CAS: 1233921-75-9 Molecular Formula: C813CH10F215N2O5 Molecular Weight (g/mol): 267.16 Synonym: 2’-Deoxy-2’,2’-difluorouridine-13C,15N2,2',2'-Difluorodeoxyuridine-13C,15N2; SMILES: O[C@H]1C(F)(F)[C@H]([15N]2[13C]([15NH]C(C=C2)=O)=O)O[C@@H]1CO