accessibility menu, dialog, popup

UserName

H2NNH2

CAS RN 124-09-4

IUPAC Name: hexane-1,6-diamine
Synonyms: 1,6-diaminohexane hmda cpd HMDA 1,6-hexanediamine (solution) hex-NH2
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: NAQMVNRVTILPCV-UHFFFAOYSA-N
SMILES: NCCCCCCN
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (6)
Boiling Point 
  • (3)
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)