accessibility menu, dialog, popup

UserName

SFFFFFClOH

CAS RN 1240256-97-6

IUPAC Name: [3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Synonyms: [3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Molecular Weight (g/mol): 268.63
Molecular Formula: C7H6ClF5OS
InChi Key: JXKDNNYDGFDTFO-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results