CAS RN 1240257-20-8
IUPAC Name:
1-(bromomethyl)-3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms:
1-(bromomethyl)-3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol):
315.07
Molecular Formula:
C7H5BrF6S
InChi Key:
WKTXJYLDFXWFOO-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC(CBr)=C1)S(F)(F)(F)(F)F
1
–
1
of
1
results
1
3-Fluoro-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™
CAS: 1240257-20-8 Molecular Formula: C7H5BrF6S Molecular Weight (g/mol): 315.07 MDL Number: MFCD16652459 InChI Key: WKTXJYLDFXWFOO-UHFFFAOYSA-N Synonym: 3-fluoro-5-pentafluorosulfur benzyl bromide,1-bromomethyl-3-fluoro-5-pentafluoro-??-sulfanyl benzene,3-fluoro-5-pentafluorothio benzyl bromide PubChem CID: 66523538 IUPAC Name: [3-(bromomethyl)-5-fluorophenyl]-pentafluoro-$l^{6}-sulfane SMILES: FC1=CC(=CC(CBr)=C1)S(F)(F)(F)(F)F