accessibility menu, dialog, popup

UserName

FSFFFFFOHO

CAS RN 1240257-84-4

IUPAC Name: 2-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Synonyms: [3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Molecular Weight (g/mol): 280.18
Molecular Formula: C8H6F6O2S
InChi Key: IZLGUBFCHOPNJB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results