accessibility menu, dialog, popup

UserName

BNNFFFK

CAS RN 1242733-91-0

IUPAC Name: potassium trifluoro(pyrimidin-5-yl)boranuide
Synonyms: potassium trifluoro(pyrimidin-5-yl)boranuide
Molecular Weight (g/mol): 185.99
Molecular Formula: C4H3BF3KN2
InChi Key: FLVBHQQPIJARSC-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results