missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium pyrimidine-5-trifluoroborate, 95%

Product Code. 15414567
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414567 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414567 Supplier Alfa Aesar™ Supplier No. H31906.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1242733-91-0
Molecular Formula C4H3BF3KN2
Molecular Weight (g/mol) 185.986
MDL Number MFCD09992922
InChI Key FLVBHQQPIJARSC-UHFFFAOYSA-N
Synonym potassium pyrimidine-5-trifluoroborate, potassium trifluoro pyrimidin-5-yl boranuide, potassium trifluoro pyrimidin-5-yl borate, pyrimidine-5-trifluoroborate potassium salt, potassium trifluoro pyrimidin-5-yl borate 1-
PubChem CID 45588203
IUPAC Name potassium;trifluoro(pyrimidin-5-yl)boranuide
SMILES [B-](C1=CN=CN=C1)(F)(F)F.[K+]

Specifications

Quantity 250 mg
Formula Weight 185.99
Percent Purity 95%
Chemical Name or Material Potassium pyrimidine-5-trifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.