accessibility menu, dialog, popup

UserName

CAS RN 12607-70-4

H2OH2OH2OH2ONiNiNiOOO

CAS RN 12607-70-4

IUPAC Name: trinickel(2+) tetrakis(λ³-oxidanide) carbonate
Synonyms: trinickel(2+) tetrakis(λ³-oxidanide) carbonate
Molecular Weight (g/mol): 308.15
Molecular Formula: CH8Ni3O7
InChi Key: JYTXCCGDBBXJAQ-UHFFFAOYSA-L
SMILES: [OH2-].[OH2-].[OH2-].[OH2-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results