accessibility menu, dialog, popup

UserName

CAS RN 1261673-90-8

ClOFFFOH

CAS RN 1261673-90-8

IUPAC Name: [4-chloro-2-(trifluoromethoxy)phenyl]methanol
Synonyms: [4-chloro-2-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 226.58
Molecular Formula: C8H6ClF3O2
InChi Key: DAXPTNWTUUAZSS-UHFFFAOYSA-N
SMILES: OCC1=C(OC(F)(F)F)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results