accessibility menu, dialog, popup

UserName

CH3NNClNH2

CAS RN 126417-82-1

IUPAC Name: 3-(4-chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Synonyms: 5-(4-chlorophenyl)-2-methylpyrazol-3-amine
Molecular Weight (g/mol): 207.66
Molecular Formula: C10H10ClN3
InChi Key: ZSFCXTGPGSAWFT-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1N)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results