missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine, ≥97%, Thermo Scientific™

Product Code. 10575063
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10575063 1 g 1g
10432393 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10575063 Supplier Thermo Scientific Maybridge Supplier No. KM04038DA
 Restricted Item
Estimated Shipment: 01-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 126417-82-1
Molecular Formula C10H10ClN3
Molecular Weight (g/mol) 207.661
MDL Number MFCD00084872
InChI Key ZSFCXTGPGSAWFT-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-2-methylpyrazol-3-amine, 1h-pyrazol-5-amine,3-4-chlorophenyl-1-methyl, 5-amino-3-4-chlorophenyl-1-methyl-1h-pyrazole, 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-ylamine, 5-4-chloro-phenyl-2-methyl-2h-pyrazol-3-ylamine, maybridge1_004889, acmc-1c52v, 5-4-chlorophenyl-2-methyl-3-pyrazolamine, 5-4-chlorophenyl-2-methyl-pyrazol-3-amine
PubChem CID 2735304
IUPAC Name 5-(4-chlorophenyl)-2-methylpyrazol-3-amine
SMILES CN1C(=CC(=N1)C2=CC=C(C=C2)Cl)N

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber glass bottle
Assay Percent Range 97%
Formula Weight 207.66
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.