accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNOOH3COO

CAS RN 126937-42-6

IUPAC Name: 1-benzyl 4-tert-butyl 2-methyl piperazine-1,2,4-tricarboxylate
Synonyms: 1-benzyl 4-tert-butyl 2-methyl piperazine-1,2,4-tricarboxylate
Molecular Weight (g/mol): 378.43
Molecular Formula: C19H26N2O6
InChi Key: FUMOVDZJNXKHJM-UHFFFAOYNA-N
SMILES: COC(=O)C1CN(CCN1C(=O)OCC1=CC=CC=C1)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)