missing translation for 'onlineSavingsMsg'
Learn More

Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate, Thermo Scientific™

Product Code. 15427735
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427735 250 mg 250mg
11307419 1 g 1g
11317419 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15427735 Supplier Thermo Scientific Alfa Aesar Supplier No. H27832.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126937-42-6
Molecular Formula C19H26N2O6
Molecular Weight (g/mol) 378.425
MDL Number MFCD03426296
InChI Key FUMOVDZJNXKHJM-UHFFFAOYSA-N
PubChem CID 4451965
IUPAC Name 1-O-benzyl 4-O-tert-butyl 2-O-methyl piperazine-1,2,4-tricarboxylate
SMILES CC(C)(C)OC(=O)N1CCN(C(C1)C(=O)OC)C(=O)OCC2=CC=CC=C2

Specifications

Quantity 250 mg
Synonym 1-benzyl 4-tert-butyl 2-methyl piperazine-1,2,4-tricarboxylate, methyl n-4-boc-n-1-cbz-2-piperazinecarboxylate, methyl 4-boc-1-cbz-2-piperazinecarboxylate, methyl 4-n-boc-1-n-cbz-2-piperazine carboxylate, 4-n-boc-1-n-cbz-piperazine-2-carboxylic acid methyl ester, methyl +/--4-n-boc-1-n-benyloxycarbonyl-2-piperazinecarboxylate, +/--1-benzyloxycarbonyl-4-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,2,4-tricarboxylic acid 1-benzyl ester 4-tert-butyl ester 2-methyl ester, methyl4-boc-1-cbz-2-piperazinecarboxylate, 2-methyl 4-tert-butyl 1-benzyl piperazine-1,2,4-tricarboxylate
Formula Weight 378.43
Chemical Name or Material Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.