accessibility menu, dialog, popup

UserName

FFFSClOO

CAS RN 127162-96-3

IUPAC Name: [3-(trifluoromethyl)phenyl]methanesulfonyl chloride
Synonyms: [3-(trifluoromethyl)phenyl]methanesulfonyl chloride
Molecular Weight (g/mol): 258.64
Molecular Formula: C8H6ClF3O2S
InChi Key: WIGCBGFBBRJTLS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CS(Cl)(=O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results