accessibility menu, dialog, popup

UserName

CAS RN 127371-50-0

IUPAC Name: 1,2-difluoro-4-methyl-5-nitrobenzene
Synonyms: 1,2-difluoro-4-methyl-5-nitrobenzene
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: DJEBTFSOEQWELL-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(F)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results