accessibility menu, dialog, popup

UserName

NH2OOFFF

CAS RN 127979-74-2

IUPAC Name: 2-(trifluoromethoxy)benzamide
Synonyms: 2-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: XXTXDVUAHROLBN-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results