CAS RN 129136-65-8
IUPAC Name:
4-[(thiophen-2-yl)methyl]aniline
Synonyms:
4-(thiophen-2-ylmethyl)aniline
Molecular Weight (g/mol):
189.28
Molecular Formula:
C11H11NS
InChi Key:
PIJZLXNJWCKMSA-UHFFFAOYSA-N
SMILES:
NC1=CC=C(CC2=CC=CS2)C=C1
1
–
1
of
1
results
1
4-(thien-2-ylmethyl)aniline, Thermo Scientific™
CAS: 129136-65-8 Molecular Formula: C11H11NS Molecular Weight (g/mol): 189.276 InChI Key: PIJZLXNJWCKMSA-UHFFFAOYSA-N Synonym: 4-thien-2-ylmethyl aniline,4-thiophen-2-ylmethyl aniline,4-thiophen-2-yl methyl aniline,4-2-thienylmethyl phenylamine PubChem CID: 20063869 IUPAC Name: 4-(thiophen-2-ylmethyl)aniline SMILES: C1=CSC(=C1)CC2=CC=C(C=C2)N