missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)aniline, Thermo Scientific™

Product Code. 10707505
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10707505 1 g Amber Glass Bottle 1g
10275474 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707505 Supplier Thermo Scientific Maybridge Supplier No. CC70614DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 129136-65-8
Molecular Formula C11H11NS
Molecular Weight (g/mol) 189.276
InChI Key PIJZLXNJWCKMSA-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
PubChem CID 20063869
IUPAC Name 4-(thiophen-2-ylmethyl)aniline
SMILES C1=CSC(=C1)CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.28
Percent Purity 95%
Chemical Name or Material 4-(Thien-2-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.