accessibility menu, dialog, popup

UserName

OHBHONSOO

CAS RN 129271-98-3

IUPAC Name: [1-(benzenesulfonyl)-1H-indol-3-yl]boronic acid
Synonyms: 1-(benzenesulfonyl)indol-3-ylboronic acid
Molecular Weight (g/mol): 301.12
Molecular Formula: C14H12BNO4S
InChi Key: YKTZLHLBQGCFQX-UHFFFAOYSA-N
SMILES: OB(O)C1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results