CAS RN 129660-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Acetyl Glyphosate, TRC
CAS: 129660-96-4 Molecular Formula: C5 H10 N O6 P Molecular Weight (g/mol): 211.11 Synonym: Glycine, N-acetyl-N-(phosphonomethyl)-,N-Acetyl-N-(phosphonomethyl)glycine,N-Acetylglyphosate IUPAC Name: 2-[acetyl(phosphonomethyl)amino]acetic acid SMILES: CC(=O)N(CC(=O)O)CP(=O)(O)O