accessibility menu, dialog, popup

UserName

H3CNOPOCH3

CAS RN 129986-67-0

IUPAC Name: N-methoxy-N-methyl-2-(triphenyl-λ⁵-phosphanylidene)acetamide
Synonyms: N-methoxy-N-methyl-2-(triphenyl-λ⁵-phosphanylidene)acetamide
Molecular Weight (g/mol): 363.4
Molecular Formula: C22H22NO2P
InChi Key: DGBLSOOPDBHHRX-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results