accessibility menu, dialog, popup

UserName

OHO

CAS RN 13020-57-0

IUPAC Name: 3-benzoylphenol
Synonyms: P-hydroxybenzophenone
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: SHULEACXTONYPS-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results