accessibility menu, dialog, popup

UserName

CAS RN 13041-62-8

IUPAC Name: 4-methoxynaphthalene-1-carboxylate
Synonyms: 4-methoxynaphthalene-1-carboxylate
Molecular Weight (g/mol): 201.2
Molecular Formula: C12H9O3
InChi Key: WRQHSQDGYYDRMX-UHFFFAOYSA-M
SMILES: COC1=C2C=CC=CC2=C(C=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)