accessibility menu, dialog, popup

UserName

CAS RN 131-89-5

IUPAC Name: 2-cyclohexyl-4,6-dinitrophenol
Synonyms: 2-cyclohexyl-4,6-dinitrophenol
Molecular Weight (g/mol): 266.25
Molecular Formula: C12H14N2O5
InChi Key: QJYHUJAGJUHXJN-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results