accessibility menu, dialog, popup

UserName

CCu

CAS RN 13146-23-1

IUPAC Name: λ¹-copper(1+) phenylethyne
Synonyms: λ¹-copper(1+) phenylethyne
Molecular Weight (g/mol): 164.67
Molecular Formula: C8H5Cu
InChi Key: SFLVZUJLUPAJNE-UHFFFAOYSA-N
SMILES: [Cu+].[C-]#CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)