missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) phenylacetylide, Thermo Scientific Chemicals

Product Code. 15471077
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471077 500 mg 500mg
11303377 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471077 Supplier Thermo Scientific Alfa Aesar Supplier No. 017538.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Copper(I) phenylacetylide participates in efficient coupling with a range of aryl iodides and bromides to give substituted tolanes which, if possessing an ortho amino or hydroxy group, can be thermolyzed to yield 2-phenylbenzofurans, -indoles, and -furo[3,2-b]pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13146-23-1
Molecular Formula C8H5Cu
Molecular Weight (g/mol) 164.674
MDL Number MFCD00015578
InChI Key SFLVZUJLUPAJNE-UHFFFAOYSA-N
Synonym copper 1+ ; ethynylbenzene, copper i phenylacetylide, copper i phenylacetylide, min
PubChem CID 6101206
IUPAC Name copper(1+);ethynylbenzene
SMILES [C-]#CC1=CC=CC=C1.[Cu+]

Specifications

Color Yellow
Quantity 500 mg
UN Number UN3088
Sensitivity Air & Moisture Sensitive
Solubility Information Soluble in acetone and acetonitrile; Insoluble in water
Formula Weight 153.68
Physical Form Powder
Chemical Name or Material Copper(I) phenylacetylide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.