CAS RN 132741-29-8
IUPAC Name:
2-(3,4-difluorophenyl)-2-hydroxyacetic acid
Synonyms:
(3,4-difluorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol):
188.13
Molecular Formula:
C8H6F2O3
InChi Key:
BKHXODARAOCNDJ-UHFFFAOYNA-N
SMILES:
OC(C(O)=O)C1=CC(F)=C(F)C=C1
1
–
1
of
1
results
1
3,4-Difluoromandelic acid, 97%, Thermo Scientific™
CAS: 132741-29-8 Molecular Formula: C8H6F2O3 Molecular Weight (g/mol): 188.13 MDL Number: MFCD00061299 InChI Key: BKHXODARAOCNDJ-UHFFFAOYSA-N Synonym: 3,4-difluoromandelic acid,2-3,4-difluorophenyl-2-hydroxyacetic acid,3,4-difluorophenyl hydroxy acetic acid,difluoromandelicacid,3,4-difluoro mandelic acid,3,4-difluorophenyl hydroxy acetic acid #,alpha-hydroxy-3,4-difluorobenzeneacetic acid,2-3,4-difluorophenyl-2-hydroxy-acetic acid,2-3,4-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid PubChem CID: 588647 IUPAC Name: 2-(3,4-difluorophenyl)-2-hydroxyacetic acid SMILES: C1=CC(=C(C=C1C(C(=O)O)O)F)F