accessibility menu, dialog, popup

UserName

CH3H3CCH3OO(S)HNOOH3CH3CCH3NH2HCl

CAS RN 13288-57-8

IUPAC Name: tert-butyl 2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Synonyms: tert-butyl 2-amino-6-[(tert-butoxycarbonyl)amino]hexanoate hydrochloride
Molecular Weight (g/mol): 338.87
Molecular Formula: C15H31ClN2O4
InChi Key: TZBPQINFXPIRBX-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)NCCCCC(N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)