missing translation for 'onlineSavingsMsg'
Learn More

H-Lys(Boc)-OtBu Hydrochloric Acid Salt, TRC

Product Code. 30475421
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475421 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475421 Supplier Toronto Research Chemicals Supplier No. TRCL487900500MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13288-57-8
Molecular Formula C15H31ClN2O4
Synonym Nepsilon-Boc-L-lysine tert-butyl ester Hydrochloride, Ne-Boc-L-lysine tert-butyl Ester Hydrochloride, N6-[(1,1-Dimethylethoxy)carbonyl]-L-lysine 1,1-Dimethylethyl Ester Hydrochloride, (S)-2-Amino-6-[(tert-butoxycarbonyl)amino]hexanoic Acid tert-Butyl Ester Hydrochloride, NSC 164053, N’-tert-Butoxycarbonyl-L-lysine tert-Butyl Ester Hydrochloride, tert-Butyl (2S)-2-Amino-6-[(tert-butoxycarbonyl)amino]hexanoate Hydrochloride
InChI Formula InChI=1S/C15H30N2O4.ClH/c1-14(2,3)20-12(18)11(16)9-7-8-10-17-13(19)21-15(4,5)6;/h11H,7-10,16H2,1-6H3,(H,17,19);1H/t11-;/m0./s1
SMILES Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
Molecular Weight (g/mol) 338.87
Quantity 500 mg
Formula Weight 338.1972
Shipping Condition Room Temperature
Chemical Name or Material H-Lys(Boc)-OtBu Hydrochloric Acid Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.