accessibility menu, dialog, popup

UserName

NH2ClSHOOO

CAS RN 133-74-4

IUPAC Name: 2-amino-5-chlorobenzene-1-sulfonic acid
Synonyms: 2-amino-5-chlorobenzenesulfonic acid
Molecular Weight (g/mol): 207.63
Molecular Formula: C6H6ClNO3S
InChi Key: ZCGVPUAAMCMLTM-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results