CAS RN 1330266-29-9
CAS RN 1330266-29-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-6-mercaptopurine-13C2,15N, TRC
CAS: 1330266-29-9 Molecular Formula: C313C2H5N415NS Molecular Weight (g/mol): 170.17 Synonym: 2-Amino-1,7(9)-dihydro-purine-6-thione-13C2,15N,Tioguanine-13C2,15N,6-Thioguanine-13C2,15N,2-Amino-9H-purine-6-thiol-13C2,15N; IUPAC Name: 2-amino-3,7-dihydropurine-6-thione SMILES: SC1=NC(N)=N[13C]2=[13C]1[15N]=CN2