accessibility menu, dialog, popup

UserName

CH3OOClNOO

CAS RN 13324-11-3

IUPAC Name: methyl 2-chloro-4-nitrobenzoate
Synonyms: methyl 2-chloro-4-nitrobenzoate
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: PICNSXCJRMYANX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results