accessibility menu, dialog, popup

UserName

CH3OOHOH3C

CAS RN 13334-49-1

IUPAC Name: 2-(2,4-dimethylphenoxy)acetic acid
Synonyms: 2,4-dimethylphenoxyacetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: XRTZHWXWLUGOAT-UHFFFAOYSA-N
SMILES: CC1=CC=C(OCC(O)=O)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results