CAS RN 133565-43-2
IUPAC Name:
tert-butyl N-[3-(benzyloxy)-1-hydroxybutan-2-yl]carbamate
Synonyms:
tert-butyl N-[3-(benzyloxy)-1-hydroxybutan-2-yl]carbamate
Molecular Weight (g/mol):
295.38
Molecular Formula:
C16H25NO4
InChi Key:
RXDBGDZAKNELGW-UHFFFAOYNA-N
SMILES:
CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C
1
–
1
of
1
results
1
(2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol, 97%, Thermo Scientific™
CAS: 133565-43-2 Molecular Formula: C16H25NO4 Molecular Weight (g/mol): 295.38 MDL Number: MFCD00235948 InChI Key: RXDBGDZAKNELGW-UHFFFAOYNA-N Synonym: tert-butyl 2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate,boc-thr bzl-ol,boc-threoninol bzl,2r,3r-2-boc-amino-3-benzyloxy-1-butanol,tert-butyl n-2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate,ambotzbal1043,n-boc-l-thr bzl-ol,n-boc-o-benzyl-l-threoninol,n-boc-2r,3r-2-amino-3-benzyloxy-1-butanol PubChem CID: 11109205 IUPAC Name: tert-butyl N-[(2R,3R)-1-hydroxy-3-phenylmethoxybutan-2-yl]carbamate SMILES: CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C