accessibility menu, dialog, popup

UserName

NaH3CSOOO

CAS RN 13419-61-9

IUPAC Name: sodium decane-1-sulfonate
Synonyms: sodium decane-1-sulfonate
Molecular Weight (g/mol): 244.32
Molecular Formula: C10H21NaO3S
InChi Key: AIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (7)
  • (3)
  • (3)
  • (1)
  • (3)
  • (5)
  • (4)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)