missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic acid, sodium salt, 96%

Product Code. 10683763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10683763 100 g Glass bottle 100g
10673763 5 g Glass bottle 5g
10446252 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683763 Supplier Thermo Scientific Acros Supplier No. 369451000
  • ONLINE EXCLUSIVE PRICE
    650.00 €
    609.00 € / 100g
    Save 41.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1-Decanesulfonic acid, sodium salt, 96%, Quantity: 25g, Packaging: Glass bottle, White to Cream, Melting Point: >300 deg.C, Molecular Weight: 244.32, Percent Purity: 96%, Assay Percent Range: 96%, Beilstein: 04, III, 27, CAS: 13419-61-9, CAS Max %: 100.0, CAS Min %: 95.0 | CAS: 13419-61-9 | C10H21NaO3S | 244.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.32
MDL Number MFCD00007526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
Synonym sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
PubChem CID 2724181
IUPAC Name sodium;decane-1-sulfonate
SMILES CCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300°C
Color White to Cream
Loss on Drying 5% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3(CH2)9SO3Na
Quantity 100 g
Beilstein 04, III, 27
Solubility Information Solubility in water: soluble
Formula Weight 244.32
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1-Decanesulfonic acid, sodium salt, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.