accessibility menu, dialog, popup

UserName

CAS RN 13429-12-4

H2NOOHBrHO

CAS RN 13429-12-4

IUPAC Name: 4-bromo-3,5-dihydroxybenzamide
Synonyms: 4-bromo-3,5-dihydroxybenzamide
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: GCNIEGHLVCGBRA-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(O)=C(Br)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results